Assay Details
| Gene Symbol: Cox20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025511.2 | NP_079787.1 | 2 - 3 | 167 | 116 | |
| GenBank mRNA | AK009178.1 | - | 2 - 3 | 161 | 116 | |
| AK019293.1 | - | 1 - 2 | 59 | 116 | ||
| BC055277.1 | - | 2 - 3 | 150 | 5714061 | 116 | |
| BC059717.1 | - | 2 - 3 | 165 | 6543216 | 116 | |
| BY779933.1 | - | 2 - 3 | 167 | 116 | ||
More Information
Set Membership: |
|

Cox20
Genomic Map