Assay Details
| Gene Symbol: CIB4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164713.1 | NP_001158185.1 | 1 - 2 | 52 | 63 | |
| GenBank mRNA | FJ039532.1 | - | 1 - 2 | 52 | 63 | |
| KJ425421.1 | - | 1 - 2 | 51 | 63 | ||
More Information
Set Membership: |
|

CIB4
Genomic Map