Assay Details
| Gene Symbol: CDH12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027980146.1 | XP_027835947.1 | 8 - 9 | 1597 | 60 | |
| XM_027980147.1 | XP_027835948.1 | 8 - 9 | 1597 | 60 | ||
More Information
Set Membership: |
|

CDH12
Genomic Map