Assay Details
| Gene Symbol: ZNF180 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015100590.2 | XP_014956076.2 | 3 - 4 | 402 | 101 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF180| Gene Symbol: ZNF180 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015100590.2 | XP_014956076.2 | 3 - 4 | 402 | 101 | |
Set Membership: |
|