Assay Details
| Gene Symbol: C12H1orf21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012187716.3 | XP_012043106.1 | 2 - 3 | 373 | 70 | |
| XM_015099359.2 | XP_014954845.1 | 2 - 3 | 521 | 70 | ||
More Information
Set Membership: |
|

C12H1orf21
Genomic Map