Assay Details
| Gene Symbol: GPRC5C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015098972.2 | XP_014954458.2 | 4 - 5 | 1452 | 62 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

GPRC5C| Gene Symbol: GPRC5C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015098972.2 | XP_014954458.2 | 4 - 5 | 1452 | 62 | |
Set Membership: |
|