Assay Details
| Gene Symbol: PLAAT5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027959754.1 | XP_027815555.1 | 5 - 6 | 828 | 75 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

PLAAT5| Gene Symbol: PLAAT5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027959754.1 | XP_027815555.1 | 5 - 6 | 828 | 75 | |