Assay Details
| Gene Symbol: LOC101120595 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027977130.1 | XP_027832931.1 | 26 - 27 | 3355 | 85 | |
| XM_027977131.1 | XP_027832932.1 | 26 - 27 | 3355 | 85 | ||
| XM_027977132.1 | XP_027832933.1 | 26 - 27 | 3355 | 85 | ||
| XM_027977133.1 | XP_027832934.1 | 26 - 27 | 3355 | 85 | ||
More Information
Set Membership: |
|

LOC101120595
Genomic Map