Assay Details
| Gene Symbol: NIP7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004015098.4 | XP_004015147.1 | 3 - 4 | 490 | 90 | |
| XM_012190037.3 | XP_012045427.1 | 2 - 3 | 408 | 90 | ||
| XM_015100516.2 | XP_014956002.1 | 3 - 4 | 494 | 90 | ||
More Information
Set Membership: |
|

NIP7
Genomic Map