Assay Details
| Gene Symbol: RNF4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015096669.2 | XP_014952155.1 | 7 - 8 | 654 | 61 | |
| XM_015096670.2 | XP_014952156.1 | 7 - 8 | 635 | 61 | ||
| XM_027971300.1 | XP_027827101.1 | 7 - 8 | 715 | 61 | ||
More Information
Set Membership: |
|

RNF4
Genomic Map