Assay Details
| Gene Symbol: RBM48 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004007713.4 | XP_004007762.1 | 3 - 4 | 486 | 107 | |
| XM_027968442.1 | XP_027824243.1 | 2 - 3 | 902 | 107 | ||
More Information
Set Membership: |
|

RBM48
Genomic Map