Assay Details
| Gene Symbol: NANOS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015095791.2 | XP_014951277.2 | 1 | 2938 | 109 | |
| XM_027969516.1 | XP_027825317.1 | 1 | 2938 | 109 | ||
More Information
Set Membership: |
|

NANOS3
Genomic Map