Assay Details
| Gene Symbol: JUN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198947.1 | NP_001185876.1 | 1 | 407 | 146 | |
| GenBank mRNA | AJ291306.1 | - | 1 | 249 | 146 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: JUN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198947.1 | NP_001185876.1 | 1 | 407 | 146 | |
| GenBank mRNA | AJ291306.1 | - | 1 | 249 | 146 | |
Set Membership: |
|