Assay Details
| Gene Symbol: VDAC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082076.1 | NP_001075545.1 | 653 | 77 | ||
| XM_008273902.2 | XP_008272124.1 | 691 | 77 | |||
| GenBank mRNA | AF209727.1 | - | 653 | 77 | ||
More Information
Set Membership: |
|

VDAC3
Genomic Map