Assay Details
| Gene Symbol: RAB25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008264328.2 | XP_008262550.1 | 3 - 4 | 554 | 104 | |
| GenBank mRNA | L03303.2 | - | 2 - 3 | 339 | 104 | |
More Information
Set Membership: |
|

RAB25
Genomic Map