Assay Details
| Gene Symbol: NAA10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001171367.1 | NP_001164838.1 | 1 | 352 | 67 | |
| XM_008250281.2 | XP_008248503.1 | 581 | 67 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

NAA10| Gene Symbol: NAA10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001171367.1 | NP_001164838.1 | 1 | 352 | 67 | |
| XM_008250281.2 | XP_008248503.1 | 581 | 67 | |||
Set Membership: |
|