Assay Details
| Gene Symbol: CCL7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001297482.1 | NP_001284411.1 | 1 - 2 | 133 | 67 | |
| GenBank mRNA | JX000252.1 | - | 1 - 2 | 133 | 67 | |
More Information
Set Membership: |
|

CCL7
Genomic Map