Assay Details
| Gene Symbol: NRL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008269398.2 | XP_008267620.1 | 1 - 2 | 1925 | 105 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008269398.2 transcripts.
Set Membership: |
|

NRL
Genomic Map