Assay Details
| Gene Symbol: C9H3orf84 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008260646.2 | XP_008258868.1 | 1 - 2 | 208 | 67 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008260646.2 transcripts.
Set Membership: |
|

C9H3orf84
Genomic Map