Assay Details
| Gene Symbol: DPY19L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008261628.2 | XP_008259850.1 | 4 - 5 | 405 | 63 | |
| XM_017344422.1 | XP_017199911.1 | 4 - 5 | 405 | 63 | ||
More Information
Set Membership: |
|

DPY19L1
Genomic Map