Assay Details
| Gene Symbol: C19H17orf49 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008270822.2 | XP_008269044.1 | 4 - 5 | 534 | 80 | |
| XM_008270824.2 | XP_008269046.1 | 3 - 4 | 433 | 80 | ||
| XM_017349025.1 | XP_017204514.1 | 1 - 2 | 750 | 80 | ||
| XM_017349026.1 | XP_017204515.1 | 1 - 2 | 750 | 80 | ||
More Information
Set Membership: |
|

C19H17orf49
Genomic Map