Assay Details
| Gene Symbol: CPB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008259991.2 | XP_008258213.1 | 10 - 11 | 1135 | 61 | |
| XM_017343486.1 | XP_017198975.1 | 9 - 10 | 1010 | 61 | ||
More Information
Set Membership: |
|

CPB2
Genomic Map