Assay Details
| Gene Symbol: FGG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002716891.3 | XP_002716937.1 | 2 - 3 | 180 | 71 | |
| XM_008267306.2 | XP_008265528.1 | 2 - 3 | 180 | 71 | ||
More Information
Set Membership: |
|

FGG
Genomic Map