Assay Details
| Gene Symbol: PDE4DIPP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008264566.2 | XP_008262788.2 | 1 - 2 | 290 | 89 | |
| XM_008264567.2 | XP_008262789.2 | 1 - 2 | 289 | 89 | ||
| XM_008264569.2 | XP_008262791.2 | 1 - 2 | 290 | 89 | ||
| XM_008264570.2 | XP_008262792.2 | 1 - 2 | 290 | 89 | ||
| XM_008264576.2 | XP_008262798.2 | 1 - 2 | 289 | 89 | ||
| XM_017345940.1 | XP_017201429.1 | 1 - 2 | 291 | 89 | ||
| XM_017345945.1 | XP_017201434.1 | 1 - 2 | 291 | 89 | ||
More Information
Set Membership: |
|

PDE4DIPP2
Genomic Map