Assay Details
| Gene Symbol: LOC4332482 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015773501.2 | XP_015628987.1 | 2 - 3 | 197 | 79 | |
| GenBank mRNA | AK100125.1 | - | 2 - 3 | 185 | 79 | |
| KY170874.1 | - | 2 - 3 | 49 | 79 | ||
More Information
Set Membership: |
|

LOC4332482
Genomic Map