Assay Details
| Gene Symbol: LOC4336981 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015780217.2 | XP_015635703.1 | 4 - 5 | 523 | 121 | |
| XM_015780218.2 | XP_015635704.1 | 4 - 5 | 1607 | 121 | ||
| XM_015780219.2 | XP_015635705.1 | 3 - 4 | 472 | 121 | ||
| GenBank mRNA | AY177698.1 | - | 3 - 4 | 422 | 121 | |
More Information
Set Membership: |
|

LOC4336981
Genomic Map