Assay Details
| Gene Symbol: LOC4343003 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015792198.2 | XP_015647684.1 | 11 - 12 | 1280 | 102 | |
| XM_015792199.1 | XP_015647685.1 | 11 - 12 | 1257 | 102 | ||
| GenBank mRNA | AK070846.1 | - | 11 - 12 | 1259 | 102 | |
More Information
Set Membership: |
|

LOC4343003
Genomic Map