Assay Details
| Gene Symbol: LOC4345404 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015795076.2 | XP_015650562.1 | 1 - 2 | 191 | 67 | |
| GenBank mRNA | AK071493.1 | - | 1 - 2 | 191 | 67 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015795076.2 transcripts.
Set Membership: |
|

LOC4345404
Genomic Map