Assay Details
| Gene Symbol: LOC4335342 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015781016.2 | XP_015636502.1 | 11 - 12 | 1066 | 116 | |
| XM_015781018.2 | XP_015636504.1 | 11 - 12 | 965 | 116 | ||
| XM_015781019.2 | XP_015636505.1 | 9 - 10 | 751 | 116 | ||
| XM_015781021.2 | XP_015636507.1 | 10 - 11 | 831 | 116 | ||
More Information
Set Membership: |
|

LOC4335342
Genomic Map