Assay Details
| Gene Symbol: LOC4335342 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015781016.2 | XP_015636502.1 | 8 - 9 | 835 | 91 | |
| XM_015781018.2 | XP_015636504.1 | 8 - 9 | 734 | 91 | ||
| XM_015781019.2 | XP_015636505.1 | 6 - 7 | 520 | 91 | ||
| XM_015781021.2 | XP_015636507.1 | 7 - 8 | 600 | 91 | ||
| GenBank mRNA | AK069000.1 | - | 8 - 9 | 765 | 91 | |
More Information
Set Membership: |
|

LOC4335342
Genomic Map