Assay Details
| Gene Symbol: LOC4335790 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015778048.2 | XP_015633534.1 | 4 - 5 | 1299 | 98 | |
| GenBank mRNA | AK072276.1 | - | 4 - 5 | 1306 | 98 | |
| AY578159.1 | - | 4 - 5 | 1299 | 98 | ||
| EU095553.1 | - | 4 - 5 | 1254 | 98 | ||
More Information
Set Membership: |
|

LOC4335790
Genomic Map