Assay Details
| Gene Symbol: LOC4328747 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015771617.2 | XP_015627103.1 | 3 - 4 | 514 | 131 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC4328747| Gene Symbol: LOC4328747 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015771617.2 | XP_015627103.1 | 3 - 4 | 514 | 131 | |
Set Membership: |
|