Assay Details
| Gene Symbol: LOC4350987 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015762363.2 | XP_015617849.1 | 9 - 10 | 1121 | 103 | |
| GenBank mRNA | AK242785.1 | - | 9 - 10 | 1122 | 103 | |
More Information
Set Membership: |
|

LOC4350987
Genomic Map