Assay Details
| Gene Symbol: LOC4329696 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015769671.2 | XP_015625157.1 | 10 - 11 | 1515 | 109 | |
| XM_015769672.2 | XP_015625158.1 | 9 - 10 | 660 | 109 | ||
| GenBank mRNA | AK243408.1 | - | 10 - 11 | 1481 | 109 | |
More Information
Set Membership: |
|

LOC4329696
Genomic Map