Assay Details
| Gene Symbol: LOC4332612 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015775486.2 | XP_015630972.1 | 14 - 15 | 2501 | 131 | |
| XM_015775487.2 | XP_015630973.1 | 12 - 13 | 1930 | 131 | ||
| GenBank mRNA | AK243493.1 | - | 14 - 15 | 2346 | 131 | |
More Information
Set Membership: |
|

LOC4332612
Genomic Map