Assay Details
| Gene Symbol: LOC4332876 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015777305.2 | XP_015632791.1 | 3 - 4 | 295 | 93 | |
| GenBank mRNA | AJ270955.1 | - | 3 - 4 | 258 | 93 | |
| AK073938.1 | - | 3 - 4 | 281 | 93 | ||
More Information
Set Membership: |
|

LOC4332876
Genomic Map