Assay Details
| Gene Symbol: LOC4342538 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791204.2 | XP_015646690.1 | 2 - 3 | 364 | 74 | |
| GenBank mRNA | AK065897.1 | - | 2 - 3 | 366 | 74 | |
More Information
Set Membership: |
|

LOC4342538
Genomic Map