Assay Details
| Gene Symbol: LOC4346520 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015795681.2 | XP_015651167.1 | 13 - 14 | 1379 | 65 | |
| GenBank mRNA | AK059654.1 | - | 11 - 12 | 1080 | 65 | |
| AK102536.1 | - | 13 - 14 | 1254 | 65 | ||
More Information
Set Membership: |
|

LOC4346520
Genomic Map