Assay Details
| Gene Symbol: LOC4351605 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015764368.2 | XP_015619854.1 | 2 - 3 | 503 | 73 | |
| GenBank mRNA | AK065623.1 | - | 2 - 3 | 482 | 73 | |
| AK072953.1 | - | 2 - 3 | 313 | 73 | ||
More Information
Set Membership: |
|

LOC4351605
Genomic Map