Assay Details
| Gene Symbol: LOC4333841 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015775411.1 | XP_015630897.1 | 10 - 11 | 1457 | 63 | |
| GenBank mRNA | AK059920.1 | - | 6 - 7 | 785 | 63 | |
| AK100165.1 | - | 10 - 11 | 1344 | 63 | ||
| AK103027.1 | - | 10 - 11 | 1340 | 63 | ||
| AK105026.2 | - | 10 - 11 | 1341 | 63 | ||
| KF214527.1 | - | 10 - 11 | 1334 | 63 | ||
| KM982771.1 | - | 10 - 11 | 1316 | 63 | ||
| U28047.3 | - | 10 - 11 | 1349 | 63 | ||
More Information
Set Membership: |
|

LOC4333841
Genomic Map