Assay Details
| Gene Symbol: LOC4332017 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015777401.2 | XP_015632887.1 | 5 - 6 | 962 | 104 | |
| GenBank mRNA | AK121902.1 | - | 5 - 6 | 959 | 104 | |
More Information
Set Membership: |
|

LOC4332017
Genomic Map