Assay Details
| Gene Symbol: LOC4324344 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015782835.2 | XP_015638321.1 | 1 | 753 | 78 | |
| GenBank mRNA | AK103489.1 | - | 1 | 742 | 78 | |
| AK105463.1 | - | 1 | 753 | 78 | ||
More Information
Set Membership: |
|

LOC4324344
Genomic Map