Assay Details
| Gene Symbol: LOC9267485 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015782797.2 | XP_015638283.1 | 14 - 15 | 1295 | 104 | |
| XM_015782799.2 | XP_015638285.1 | 14 - 15 | 1275 | 104 | ||
| GenBank mRNA | AK073973.1 | - | 9 - 10 | 2234 | 104 | |
More Information
Set Membership: |
|

LOC9267485
Genomic Map