Assay Details
| Gene Symbol: LOC4348496 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015759277.2 | XP_015614763.1 | 11 - 12 | 1507 | 69 | |
| GenBank mRNA | AK066800.1 | - | 11 - 12 | 1505 | 69 | |
| AK099380.1 | - | 11 - 12 | 1406 | 69 | ||
| AK104008.1 | - | 7 - 8 | 813 | 69 | ||
More Information
Set Membership: |
|

LOC4348496
Genomic Map