Assay Details
| Gene Symbol: LOC4332904 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015777208.2 | XP_015632694.1 | 3 - 4 | 511 | 99 | |
| GenBank mRNA | AK066313.1 | - | 3 - 4 | 509 | 99 | |
| AK066352.1 | - | 3 - 4 | 463 | 99 | ||
More Information
Set Membership: |
|

LOC4332904
Genomic Map