Assay Details
| Gene Symbol: LOC4323940 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015766860.2 | XP_015622346.1 | 11 - 12 | 1576 | 76 | |
| XM_015766861.2 | XP_015622347.1 | 11 - 12 | 1576 | 76 | ||
More Information
Set Membership: |
|

LOC4323940
Genomic Map