Assay Details
| Gene Symbol: LOC4324732 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015778213.2 | XP_015633699.1 | 2 - 3 | 1806 | 78 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC4324732| Gene Symbol: LOC4324732 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015778213.2 | XP_015633699.1 | 2 - 3 | 1806 | 78 | |
Set Membership: |
|