Assay Details
| Gene Symbol: LOC4325106 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015794675.2 | XP_015650161.1 | 11 - 12 | 1193 | 130 | |
| XM_015794687.2 | XP_015650173.1 | 10 - 11 | 1156 | 94 | ||
| GenBank mRNA | AK243365.1 | - | 10 - 11 | 1146 | 94 | |
More Information
Set Membership: |
|

LOC4325106
Genomic Map