Assay Details
| Gene Symbol: LOC4337172 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015781193.1 | XP_015636679.1 | 3 - 4 | 1582 | 97 | |
| XM_015781194.2 | XP_015636680.1 | 4 - 5 | 688 | 97 | ||
| XM_015781195.2 | XP_015636681.1 | 4 - 5 | 682 | 97 | ||
| GenBank mRNA | AK059892.1 | - | 1 - 2 | 123 | 97 | |
| AK062620.1 | - | 4 - 5 | 424 | 97 | ||
| AK109401.2 | - | 4 - 5 | 420 | 97 | ||
More Information
Set Membership: |
|

LOC4337172
Genomic Map