Assay Details
| Gene Symbol: LOC4344227 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015789626.2 | XP_015645112.1 | 2 - 3 | 523 | 58 | |
| XM_015789627.2 | XP_015645113.1 | 2 - 3 | 166 | 58 | ||
| GenBank mRNA | AK072772.1 | - | 1 - 2 | 1028 | 58 | |
| AK108593.1 | - | 2 - 3 | 323 | 58 | ||
More Information
Set Membership: |
|

LOC4344227
Genomic Map